New Pipette Controller with Ergonomic Design
2/23/2018
Omega Zen™ Pipette Controller Provides Exceptional Control in an Ergonomic Design
February 23, 2018, Vernon Hills, Ill.— Get over nine hours of continuous and accurate use with the Argos Technologies® Omega Zen pipette controller. This portable, lightweight, and ergonomic pipette controller can be used with plastic or glass pipettes from 1 to 100 mL. The pipette controller’s balanced design and contoured buttons provides comfort even after hours of use.
Additional features include an easy-to-read liquid crystal display (LCD) that shows aspirate/dispense speed, gravity (drop) mode and battery charge. Aspirate/dispense speed can be finely tuned using thumbwheel and triggers. Lithium-ion battery charge status is accurately reflected on the LCD. The included auto-charging bench stand provides quick battery charging.
VapLock Closed Systems are modular and versatile, with a full range of available options to adapt to most any equipment setup including variety of materials for chemical compatibility and multiple port connections and bottle cap sizes.
For more information on the Omega Zen pipette controller, go to coleparmer.com or call 1-800-323-4340.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Ismatec®, Oakton®, Digi-Sense™, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 1-800-323-4340. International customers, call 1-847-549-7600. You can also visit www.coleparmer.com.