Vernon Hills, IL—Cole-Parmer is helping customers save hours of time in the laboratory and out in the field with its IoT. The company offers secure and reliable Traceable® data loggers, Jenway® spectrophotometers and Masterflex® peristaltic pumps compatible with a cloud-enabled service on certain models. These systems provide a secure cloud connection using proprietary LIVE implementation, ensuring that research and day-to-day work is well protected. Stop by Cole-Parmer’s booth #3125 at Pittcon, March 3 to 5, 2020 at McCormick Place, Chicago to see these latest products and talk with experts about how to implement IoT into a lab or process.
TraceableLive®
Constantly monitor temperature, humidity, liquid nitrogen, CO2, and barometric pressure with a variety of TraceableLive wireless technology sensors, loggers, and other cloud-monitoring devices. This technology is suitable for monitoring critical environments and receiving on-time alerts to any temperature changes. TraceableLive securely connects to data via Wi-Fi on a smartphone, tablet, or PC. The cloud-based interface requires no additional software and connecting is simple and easy with a low-cost TraceableLive subscription; some products have free subscriptions. The TraceableLive subscription adds 21 CFR Part 11 compliance and is appropriate for laboratories working under strict regulations.
CPLive™
Access lab results and protocols with a simple touch of a button using CPLive cloud-enabled Jenway 74 single-beam and 76 split-beam spectrophotometers. Safely and securely upload and archive data to the cloud; manage multiple devices simultaneously through the CPLive app on a computer, tablet, or smartphone; share data with colleagues; and access unlimited data storage. CPLive data is date- and time-stamped, encrypted, stored, and backed up 24/7.
MasterflexLive®
Remotely control and monitor select Masterflex L/S® and I/P® pumps equipped with MasterflexLive; a secure, cloud-based platform. The pumps monitor and adjust critical processes running 24/7 from anywhere. This smart technology, a first-of-its-kind for peristaltic pumps, provides real-time control of all pump parameters including speed, flow rate and dispense volume. Alerts for operating conditions and error messages are provided via push notifications, SMS and e-mail. The premium MasterflexLive subscription option adds 21 CFR Part 11 and EU Annex 11 compliance. This provides extra assurance to laboratories working under strict regulations that data in the cloud is secure and that labs meet all standard requirements.
Popular Categories
- Balances and Scales
- Analytical Balances
- Bench and Floor Scale Bases
- Bench and Floor Scales
- Counting Scales
- Crane and Hanging Scales
- Health Scales
- Indicators for Scales
- Mechanical Balances
- Moisture Balances
- Platform and Pallet Scales
- Production Weighing
- Semi and Micro Analytical Balances
- Toploading Balances
- Weighing Accessories
- Weighing Dishes
- Calibration
- Electrical Products
- Batteries and Battery Testers
- Cable Testers
- Cable Ties
- Clamp Meters
- Electrical Accessories
- Electronics Enclosures
- Function Generators
- Gauss Meters
- Insulation Testers
- LCR Meters
- Megohmmeters
- Motor Rotation Testers
- Multimeters
- Oscilloscopes
- Outlet Strips
- Power Meters and Analyzers
- Power Supplies
- Resistance Testers
- RF Signal Generators
- Static Control Products
- Surge Protectors
- Test Leads and Probes
- Voltage Detectors
- Filtration Products
- Fluid Handling
- Furniture
- Heating and Cooling Equipment
- Laboratory Equipment
- Baths and Circulators
- Burettes (Burets)
- Calorimeters
- Centrifuges
- Chromatography Products
- Desiccators
- Digestion Systems
- Dispensers
- Distillation
- Electrophoresis Products
- Electroporation Products
- Environmental Samplers
- Extraction Apparatus
- Fermentation Products
- Flame Photometers
- Fluid Bed Dryers
- Freeze Dryers
- Glassware Washers
- Histology Products
- Homogenizers and Ultrasonic Processors
- Humidifiers and Dehumidifiers
- Lab Shakers
- Laboratory Evaporators
- Laboratory Sieves and Sieve Shakers
- Leachate Equipment
- Lighting and Illumination
- Magnetic Stirrers and Hot Plates
- Microscopes and Magnification
- Mixers and Blenders
- Particle Counters
- Pipettes and Pipettors
- Refractometers
- Thermal Cyclers
- Titrators
- Ultrasonic Cleaners
- Vacuums
- Water Purification Products
- Laboratory Supplies
- Air Sampling
- Animal Research Products
- Bags
- Beakers
- Books and Reference Materials
- Bottles and Jars
- Calculators
- Carboys
- Chemistry Kits
- Clean Room Stationery
- Containers
- Cuvettes
- Dialysis Products
- Dippers
- Distillation Supplies
- Field Sampling
- Flasks
- Graduated Cylinders
- Ice Buckets and Ice Pans
- Lab Cleaning Products
- Lab Organizers
- Labels and Label Printers
- Laboratory Dishes
- Laboratory Timers and Clocks
- Labware
- Microbiology Products
- Microplate Products
- Mortars and Pestles
- Parts and Accessories
- Racks
- Sampling Wipes
- Shippers
- Syringes
- Tubes
- Vials
- Watch Glasses
- Material Handling and Storage
- Material Testing and Physical Testing
- Pressure and Vacuum Instruments
- Process Controllers
- Pumps
- Safety Products
- Air Quality Monitors
- Arc Flash Products
- Combustion Analyzers
- Eyewash
- Eyewear and Safety Glasses
- Floor Mats and Matting
- Gas Detectors
- Gloves
- Hearing Protection
- Lockout and Tagout Product
- Personal Monitor Badges
- Radiation Detectors
- Refrigerant Products
- Respirators and Masks
- Safety Apparel
- Safety Shields
- Safety Signs and Labels
- Solvent Safety
- Sound Meters
- Spill Control
- Temperature Measurement Products
- Test and Measurement Products
- Water Quality Products
- Colorimeters
- Colorimetry Reagents
- Conductivity, Resistivity, and TDS Meters
- Dissolved Oxygen Meters
- Dye Tracers
- Electrodes
- Flocculation Testers
- Multiparameter Meters
- pH Buffers
- pH Meters
- Turbidity Accessories
- Turbidity Meters
- Water Quality Accessories
- Water Quality Standards, and Solutions
- Water Quality Test Strips



