Fully modular to configure as needed, with up to five parameter modules available
For an advanced electrochemistry meter that offers versatility for multiple uses, Cole-Parmer introduces the Thermo Scientific Orion VERSA STAR™ Meter. Measure up to four channels at once! Customize channels by selecting from five parameter modules: pH, pH/ISE, conductivity, DO/RDO, and pH with LogR. Read measurements on the large color graphic LCD with intuitive multilanguage (English,Spanish, French, Italian, German, and Chinese) interface. Non-volatile memory holds up to 2000 data points with time and date stamp. Transfer data via two USB and RS232 ports.
Choose the meter with specific modules only, the meter with module package, or a full kit containing meter, modules, electrodes, and solutions. For more information, call 800-323-4340 or visit ColeParmer.com/19071.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at www.coleparmer.com.
Download 300 DPI 58825_80.jpg Image