A true breakthrough in chemical instrumentation! The world’s first miniature NMR spectrometer is affordable, portable, and easy to use for multiple applications.
Mumbai, 28 February, 2011—Researchers and educators now have access to a revolutionary benchtop NMR spectrometer. The 45-MHz picoSpin spectrometer resolves proton chemical shifts with a resolution of 100 parts per billion using a very small sample volume of less than 20 micro liters. Liquid samples are simply injected into an internal capillary of the device via front-panel fittings.
“We have engineered an innovative and disruptive technology that changes the NMR spectroscopy landscape. With its low price, compact footprint and true spectroscopic capability, the picoSpin-45 spectrometer enables applications that could not be considered in the past,” said John Price, Ph.D., president and chief scientist of picoSpin, LLC.
The picoSpin-45 NMR Spectrometer is a finalist in the Science & Medical – Diagnostic Aids category, one of twelve categories honored by the Edison Awards.
The complete liquid-phase proton NMR system includes permanent magnet, transmitter, receiver, data acquisition, programmable pulse sequencer, Ethernet interfaces and intuitive web-based control software. To view this innovation, go to ColeParmer.in/8332 or, for more information, call 022-6716-2222.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 403-404, B-Wing, Delphi Hiranandani Business Park, Powai, Mumbai 400 076. In India, call 022-6716-2222. Visit us at www.coleparmer.in.
Download 300 DPI 83090_00.jpg Image