An economical unit that determines the concentration of five elements
Vernon Hills, Ill., October 20, 2011—An affordable option in flame photometers is here—the single channel, analog output, and digital display flame photometer from Cole-Parmer. Five filters for sodium, potassium, lithium, calcium, and barium are included with the unit as a standard feature. Inheriting the mixing chamber, burner stem, and burner head from a higher-cost design, this unit offers the best limits of detection available from a low-cost instrument.
The Cole-Parmer® Single-Channel Flame Photometer is ergonomically designed with a front-loading, unobstructed sample work area with removable spill containment tray. Select the filter for the element to be analyzed using the filter stick. The front panel LED indicates the element tested as well as flame status. Press “hold” to capture readings until recorded. Auto ignition and flame optimization deliver ease of use. Also includes automatic gas shutoff. With a small footprint (approx. 8” x 12”; 20 x 30 cm), the unit takes up minimal space on the lab bench. Easily clean and maintain the instrument without the requirement of tools.
For more information, call 800-323-4340 or visit ColeParmer.com/18823.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at www.coleparmer.com.
Download 300 DPI 02655_90.jpg Image