Introducing an Updated Selection of StableTemp Refrigerators and All-New Freezers from Cole-Parmer
6/27/2011
Expanded offerings include a wide range of cubic feet capacities to meet your needs
Vernon Hills, Ill., June 27, 2011—Cole-Parmer’s popular StableTemp® brand has released an updated selection of refrigerators and new freezers for scientific, medical, and pharmaceutical facilities. Each of the units offers functional features all their own, plus fully adjustable, epoxy-coated interior shelves and heavy-duty casters for easy movement. Factory-installed chart recorders are also available on select models. Choose from:
• Versatile StableTemp Laboratory Refrigerators, a robust option for general laboratory needs. These dual-hinged, single glass-door units contain industrial-grade magnetic door seals. Keep samples secure with the key lock. Choose from 12 to 72 cubic feet capacities.
StableTemp Chromatography Refrigerators, designed for laboratory research and storage for hospitals, clinics, and R&D. They feature double glass doors with industrial-grade magnetic door seals. An internal duplex outlet provides an interior electrical supply. The refrigerators offer flexible side entry access to maintain temperature continuity. Secure contents with the key lock. Select from 30 to 72 cubic feet capacities.
• All-new stainless steel, single-door StableTemp Lab Freezers are ideal for storing samples that require -12 to -30°C (10.4 to -22°F) temperatures. The freezers feature built-in alarms to alert users when the temperature moves out of range. Separate entry allows access to contents without opening the entire door. The freezers are available in 25 to 49 cubic feet capacities.
For more detailed information on these StableTemp models, call 800-323-4340 or visit ColeParmer.com/8607.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the U.S., call 800-323-4340. International customers, call 847-549-7600. Visit us at www.coleparmer.com.
Download 300 DPI 44261_44.jpg Image