“Plug-and-Play” smart meter from Cole-Parmer allows users to switch electrodes to test multiple parameters
Available for pH/ORP, conductivity, and optical DO/BOD, It’s now easier than ever to get the convenience of multi- functionality in a water quality meter. The “plug-and-play” feature of the new YSI® MultiLab DO/BOD Benchtop Meter Kits from Cole-Parmer lets users exchange sensors for optical dissolved oxygen, biochemical oxygen demand (BOD), and pH measurements, as well as operate it as a conductivity meter.
For water and wastewater applications, quality control for the food & beverage, pharmaceutical, and chemicals industries, the Smart Meter Technology offers several advantages. Manual and automatic data logging has GLP traceability and with highly accurate measurements, it displays readings in large graphics on a backlit screen. As well as that, the CMC function will continually monitor the sensors, measuring range against calibration range. The sensors digitally store their serial number, unique ID, and calibration data and transfer this data when moved.
Select from the single-channel meter, two-channel meter, or three-channel meter. To view the YSI MultiLab DO/BOD Benchtop Meter Kits , go to ColeParmer.co.uk/21388 or, for more information, call 020-8574-7543.
Download 300 DPI 05510_16.jpg Image