Cole-Parmer Introduces Sterile Mixed Cellulose Esters (MCE) Membranes for Dispensers from quality Japanese Filtration Manufacturer ADVANTEC®
The pleated band membranes offer the same high quality and reliability as individually wrapped MCE membranes and provide hands-free convenience and increased efficiency as well as reducing the risk of membrane contamination from handling. Used in many applications for the microbiological examination of water, wastewater, pharmaceuticals, food, and beverages. The membrane not only has high recovery rates, its high porosity provides superior flow rates as well.
Key benefits: -
- Available in 0.45ìm and 0.80ìm pore sizes
- A choice of membranes with gridded surface
- white (for examination of water and wastewater and applications in the pharmaceutical, medical, food and beverage industries)
- Black (for examination of bottled water, carbonated beverages, beer, and wine)
- Pre-sterilized and ready to use
- 100 individually sealed filters in a continuous pleated band
- Compatible with various dispensers (Microsart e.motion, EZ-Pak, Membrane-Butler)
- Membrane specifications, lot number, and a sequential number are printed on the clear cover of each individually wrapped filter
ADVANTEC® membranes were developed and are manufactured to comply with the ASTM and the Standard Methods for the Examination of Water and Wastewater. The filter products are manufactured to exacting standards and have established quality control specifications to assure product consistency, reproducibility, uniform performance, and superior product integrity. ADVANTEC® manufacture under a strict quality management system and are ISO 9001 approved.
Further information available on ColeParmer.co.uk/19382. To discuss your specific applications and requirements call 0500-345-300 and ask to speak to an Application Specialist.
Download 300 DPI 16644_14.jpg Image