Greater cleaning power & Enhanced Reliability with New Ultrasonic Cleaners from Cole-Parmer
3/8/2013
Overlapping sound waves thoroughly scrub even delicate materials
Using high frequency 40 kHz sound waves, the new Cole-Parmer® Ultrasonic Cleaners raise the benchmark for greater cleaning power and enhanced reliability. In addition to cleaning surgical instruments, tools, lenses, industrial parts and electronic equipment, they perform cell separation, sample preparation, and degassing liquids.
The Cole-Parmer® Ultrasonic Cleaners eliminate inconsistent cleaning with their sweep frequency which creates overlapping sound waves. The cleaners operate by transforming low-frequency AC current into 40 kHz high-frequency sound waves via a piezoelectric transducer. The transducer creates sinusoidal waves, which in turn cause cavitation — the formation and violent collapse of minute vacuum bubbles in the solution. These implosions thoroughly scrub every surface they contact, yet are not harsh on delicate items.
Choose from four models: the Cole-Parmer® Ultrasonic Cleaner with Mechanical Timer, the , the Cole-Parmer® Ultrasonic Cleaner with Digital Timer, the Cole-Parmer® Ultrasonic Cleaner with Digital Timer and Heater. To find out more about any of these models, call Cole-Parmer in the UK on 0500-345-300.
Download 300 DPI 08895_41.jpg Image