Affordable, advanced and accurate to satisfy your laboratory needs
Laboratory workers can turn to the new Cole-Parmer® Symmetry® Top-Loading and Analytical Balances to get best- performing features while still remaining budget savvy. The expanded line of weighing balances from Cole-Parmer includes:
Cole-Parmer Symmetry PA Analytical Balances offer precise weighing of small samples. The three-door glass draft shield allows easy access to the weighing chamber. A sealed keypad, splashproof design, and removable stainless steel weighing pan offer safeguards for messy lab environments.
Cole-Parmer Symmetry PT Precision Toploading Balances increase your efficiency by providing a large weighing surface for the placement of a variety of samples. Weighing modes include parts counting, percent weighing, checkweighing, animal/dynamic weighing, and density determination.
View the Cole-Parmer Symmetry Balances at Download 300 DPI 10001_02-07_10-13.jpg Image