Easy, Reliable Viscosity Measurements via Viscometer-Rheometer-on-a-Chip (VROC) Technology
8/1/2013
µVISC™ Portable Viscometers from Cole-Parmer are lightweight, fast, and accurate
Vernon Hills, Ill., August 1, 2013—Measuring viscosity is now easier than ever with portable viscometers offering simple operation yet precise results. The µVISC™ Portable Viscometers from Cole-Parmer feature a viscometer control unit, a viscosity chip, and a pipette for a complete system.
Users simply load the pipette with a sample to begin, then insert the pipette into the holder and push the run button. The microfluidic sensor measures viscosity from the drop in pressure that occurs while the sample flows through a small rectangular flow channel. The small channel size requires only a very small sample amount, reducing operational costs. The units also measure sample temperatures via the built-in RTD sensor. Users can save up to eight protocols for future reference. The previous 20 measurements, with date and time stamp, reside in memory for instant recall.
Choose from standard or advanced control units. The advanced model allows for control of the shear rate in continuous increments for greater control when measuring non-Newtonian samples. It also includes software for data analysis.
To view the µVISC™ Portable Viscometers, go to ColeParmer.com/20262 or call 800-323-4340 for more information.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at www.ColeParmer.com
Download 300 DPI 98946_40.jpg Image