New Cole-Parmer® Dual Independent Channel Touch-Screen Syringe Pump Functions Like Two Pumps
March 6, 2017, Vernon Hills, Ill — The Cole-Parmer® Dual Independent Channel Touch-Screen Syringe Pump is like getting two pumps in one. Each channel has its own motor and plunger drive, so scientists and lab technicians can program different syringe sizes, dispensing rates, and alternate directions. A large touch screen and user-friendly icon-driven software make setup quick and straightforward.
The syringe pump offers three modes of operation to satisfy life science and industrial applications: independent, reciprocating, and twin. Independent mode allows pump to function as two separate instruments with unique programming for syringe type and size, force, rate, and target volume or time. Reciprocating mode operates the two syringes in opposite directions with the same syringe type and size, and rate. Twin mode functions like traditional dual-syringe pumps using the same syringe type and size, force, rate, and target volume or time.
Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 1-800-323-4340. International customers, call 1-847-549-7600. Visit us at www.coleparmer.com.
Download jpg image