Three new models offer ultralow- to medium-level flow rates
For lab users seeking a convenient touch-screen interface, Cole-Parmer introduces three new syringe pumps. Each model uses touch-screen technology to quickly recall previous configurations or create new ones.
The Cole-Parmer® Touch-Screen Entry-Level Syringe Pumps are cost effective units that accommodate up to two syringes. They offer high precision fluid dosing with accuracy of ±0.5% and reproducibility of ±0.5%. View all operating parameters on their full-color touch screen.
The Cole-Parmer® Touch- Screen Continuous-Cycle Four-Syringe Pumps hold up to four syringes to cycle continuously back and forth in a push- pull action. As two syringes are infusing, two syringes are simultaneously withdrawing at the same rate. At the end of the set volume, the direction is automatically reversed and the next cycle begins. Offer accuracy of ±0.35% and reproducibility of ±0.5%. Daisy-chain up to 99 pumps together using the built-in RS-485.
The Cole-Parmer® Touch-Screen Nanoliter Infusion Syringe Pumps, are ideal for stereotaxic, micro manipulator, and other clamping devices. Use remote injector up to six feet away from the controller. Accurate, pulse-free flow from 3.66 pL/min to 3.818 mL/min; accuracy of ±0.5% and reproducibility of ±0.5%.
For further information and technical advice call 0500-345-300 or visit ColeParmer.co.uk/20767.
Download 300 DPI 74905_34_R.jpg Image