Easy-to-use instruments offer full-range scanning in an overall smaller size.
Vernon Hills, Ill., December 20, 2010—Reduce the time it takes to get up to speed and receive results! The Shimadzu UV/Visible Scanning Spectrophotometers feature large displays to quickly read graphical results as well as screen prompts to easily guide users. Scan over a range of 190 to 1100 nm.
Shimadzu UV/Visible Scanning Spectrophotometers are available in several models. The UVmini-1240 offers functionality from simple concentration measurements to sophisticated quantitative calibration curves. The BioSpec-mini is preprogrammed for nucleic acid quantitation and protein assays and analysis. The UV-1800 supplies data input/output and PC control via USB connectivity.
Optional accessoriesfor the UVmini-1240 model include DNA/Protein Program Pack Software, Multiwavelength Program Pack Software, Kinetics Program Pack Software, UV Data Manager Software, and Thermal Printer Kit. To learn more about the Shimadzu UV/Visible Scanning Spectrophotometers from Cole-Parmer, call 800-323-4340 or visit ColeParmer.com/8242
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the U.S., call 800-323-4340. International customers, call 847-549-7600. Visit us at www.coleparmer.com.
Download 300 DPI 83400_10.jpg Image