Ideal choice where laboratory space is limited, the new Masterflex L/S Miniflex range of economical digital peristaltic pumps from Cole-Parmer, combines accurate flow rates and advanced features in a small footprint.
These compact, stackable pumps require less than half the bench space of the L/S standard digital drive. Featuring an icon-driven menu to allow intuitive programming and set- up, the pump has a copy dispense function for filling and dispensing applications with flow rates between 0.004 and 315 mL/min. Exact flow is dependent on model and tubing size selected, but tubing calibration ensures accuracy. The single- channel model delivers the highest flow rate, while the dual- channel model provides synchronous flow from two separate channels.
Automatic tubing retention makes tube loading quick and easy while the pump head accepts continuous lengths of tubing for clean flow paths. The pump head itself features a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. The full range of Masterflex tubing formulations is available to match a variety of applications, and best in class safety and efficiency features include an open-head sensor designed to stop the drive as soon as the pump head is opened.
The durable ABS housing of the Masterflex L/S range of digital peristaltic Miniflex pumps is easy to wipe down and keep clean. Plus the self-lubricating thermoplastic polyester rollers are long wearing and low friction, ensuring a smooth operation and contributing to the long product life.
Click for information
Download 300 DPI 07525_20.jpg Image