Cole-Parmer Symmetry Balances
3/6/2017
Cole-Parmer® Symmetry® Balances are Redesigned with Both Touch and Touch-Free
March 6, 2017, Vernon Hills, Ill — Cole-Parmer® Symmetry® Touch-Screen Balances are designed with innovative features to give lab professionals more convenience and efficiency when weighing. This new family of balances provide accurate measurements of weighed loads for micro, toploading, analytical and moisture applications. An intuitive 5” capacitive touch-screen display provides clear visibility to weighing results and menu options. Infrared (IR) proximity sensors offer programmable touch-free control to minimize cross-contamination.
Advanced models are also available and equipped with extra features. Automatic and manual internal adjustments ensure accurate weighing at all times. These balances also feature external calibration, Wi-Fi connection to peripheral devices, and an “autotest GLP function” that provides an enhanced summary to aid in assessing balance operation and diagnosing reasons for occurrence of errors in weighing.
For more information, visit coleparmer.com/b/symmetry
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Ismatec®, Oakton®, Digi-Sense™, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 1-800-323-4340. International customers, call 1-847-549-7600. Visit us at www.coleparmer.com.
Download jpg image