Cole-Parmer Releases Biofuel Series of Catalogs
7/30/2012
Biodiesel, Biodiesel Blend, and Bioethanol offer a full range of products and testing information
Vernon Hills, Ill., July 30, 2012—With the release of Bioethanol, the third in a series of Biofuel catalogs from Cole-Parmer, customers have resources to choose testing equipment that helps them meet ASTM standards. Along with key testing products, the catalogs detail which specifications of ASTM apply and what is needed to pass the test methods.
The Biodiesel catalog includes critical information for meeting ASTM D6751. Biodiesel fuel blend stock (B100) is comprised of mono-alkyl esters of long chain fatty acids derived from vegetable oils or animal fats. Biodiesel fuel is normally produced from a reaction between vegetable oil or animal fat and methanol or ethanol in the presence of a catalyst to produce mono-alkyl esters and glycerin, which is removed. The biodiesel product attributes approximately 10% of its mass from the reacted alcohol. The catalog includes copper corrosion baths, carbon residue furnaces, density meters, and much more to meet testing requirements.
The Biodiesel Blend catalog references ASTM D7467 for Diesel Fuel Oil, Biodiesel Blend (B6 to B20). These fuels blend grade of 6 to 20 volume percent biodiesel with light middle or middle distillate diesel fuel. The biodiesel component of the blend must conform to the requirements of ASTM D6751, Biodiesel Fuel Blend Stock (B100) for Middle Distillate Fuels. Find a selection of equipment including analyzers, viscosity baths, oil test centrifuges, manual flash point testers, and much more.
The Bioethanol catalog contains information to meet ASTM D4806 requirements. This specification relates to nominally anhydrous denatured fuel ethanol intended to be blended with unleaded or leaded gasolines at 1 to 10% for use as automotive spark-ignition engine fuel. The denatured fuel ethanol must be visually free of sediment and suspended matter, and be clear and bright at ambient temperature or 21°C. Choose from titrators, spectrophotometers, existent gum test baths, and much more.
For more information, call 800-323-4340 or visit ColeParmer.com/19294 .
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at www.coleparmer.com.
Download 300 DPI biodfuels-bioethanol-cover.jpg Image