Cole-Parmer Presents New Thermo Scientific Steri-Cycle CO2 Incubators
11/15/2010
Achieve complete contamination control with combined HEPA air filtration and sterilization cycle to maintain cleanroom-like Class 100 air quality
Vernon Hills, Ill., November 15, 2010—Cole-Parmer offers a new option for limiting contamination in cell culture laboratories and food processing facilities. The Thermo Scientific Steri-Cycle CO2 Incubators contain an in-chamber HEPA air filtration system that filters the chamber volume every 60 seconds and reaches Class 100 air quality conditions within five minutes following a routine door opening. Plus, the incubator’s on-demand 140°C high-temperature sterilization cycle eliminates contamination from all internal surfaces— simplifying cleaning procedures.
Several programmable features make the incubators easy to operate. These include the accurate microprocessor control of temperature and CO2 concentration, the configurable audible on/off switch, access code, HEPA filter change reminder, remote alarm contacts, and low temperature and high/low CO2 alarms. The Thermo Scientific Steri-Cycle CO 2 Incubator selection includes the TC sensor model (used when temperature and relative humidity are relatively constant) and the IR sensor model (when temperature and relative humidity levels change frequently).
To view the Thermo Scientific Steri-Cycle CO2 Incubators from Cole-Parmer, visit ColeParmer.com/8182.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the U.S., call 800-323-4340. International customers, call 847-549-7600. Visit us at www.coleparmer.com.
Download 300 DPI 39320_16.jpg Image