Cole-Parmer Offers New Oakton® Temp 9000/9500 Benchtop Temperature Controllers
3/14/2013
More accurate control combined with flexible features
Vernon Hills, Ill., March 184, 2013—Temperature control just became more efficient with the new Oakton® Temp 9000/9500 Benchtop Temperature Controllers. The enhanced temperature controllers from Cole-Parmer allow users to plug heating devices right into the convenient receptacles for more accurate control.
The Oakton® Temp 9000 Thermocouple Controller offers a simultaneous view of both the actual reading and set point on the two-line alphanumeric display. The selectable temperature scale reads in °F, °C, K (Kelvin), or °R (Rankine). It is mountable on a laboratory frame or grid. The 9000 model accepts eight thermocouple types with miniconnectors.
The Oakton® Temp 9500 Advanced Multiparameter Controller builds on all of the benefits of the 9000 model, with even more enhanced features including sophisticated PID control. The higher heater output capacity and ramp-and-soak profiling is useful for more complicated processes. A USB computer interface and free software assist with real-time graphing and programming setup. The unit offers 4 to 20 mA and 1 to 5 V outputs for connection to a recorder or data logger. In addition to accepting eight thermocouple types with miniconnectors, it also accepts thermistor and RTD probes.
Both the 9000 and 9500 temperature controllers have built-in safety features that make them laboratory-friendly. They automatically shut down in the event of a broken sensor or control loop break. Their overtemperature protection shuts down the system via the user-settable limit or timer. Their outlets’ power receptacles are fused separately. They contain both audible and visual out-of-range alarms.
For more information, call 800-323-4340 or go to ColeParmer.com/19909.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at ColeParmer.com
Download 300 DPI 89000_00.jpg Image