Overlapping sound waves thoroughly scrub even delicate materials
Vernon Hills, Ill., January 14, 2013—Using high frequency 40 kHz sound waves, the new Cole-Parmer® Ultrasonic Cleaners raise the benchmark for greater cleaning power and enhanced reliability. In addition to cleaning surgical instruments, tools, lenses, industrial parts and electronic equipment, they perform cell separation, sample preparation, and degassing liquids.
The Cole-Parmer® Ultrasonic Cleaners eliminate inconsistent cleaning with their sweep frequency which creates overlapping sound waves. The cleaners operate by transforming low-frequency AC current into 40 kHz high-frequency sound waves via a piezoelectric transducer. The transducer creates sinusoidal waves, which in turn cause cavitation—the formation and violent collapse of minute vacuum bubbles in the solution. These implosions thoroughly scrub every surface they contact, yet are not harsh on delicate items.
Choose from four models: the Cole-Parmer® Ultrasonic Cleaner with Mechanical Timer , the Cole-Parmer® Ultrasonic Cleaner with Mechanical Timer and Heater, the Cole-Parmer® Ultrasonic Cleaner with Digital Timer , the Cole-Parmer® Ultrasonic Cleaner with Digital Timer and Heater. To find out more about any of these models, call 800-323-4340 or visit coleparmer.com.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at www.coleparmer.com.
Download 300 DPI 08895_43app.jpg Image