Cole-Parmer Introduces Economical Binary Gradient System for Chromatography
6/26/2014
Rugged, reliable, repeatable HPLC system even includes computer and software
Vernon Hills, Ill., June 26, 2014—It’s an ideal option for teaching and daily analytical liquid chromatography applications. The new Cole-Parmer® LC2 System is a complete high-pressure binary gradient system that performs beyond its price point. Use it for complex mixture analysis, synthesizing chemical compounds, quality control, and more.
The fully-integrated HPLC system includes a high-pressure binary pump, fixed-length UV detector, and a computer preloaded with Clarity software. The systems are available with a type 316 stainless steel or metal-free, biocompatible PEEK fluid path for chemical compatibility. The pump elicits minimal baseline noise and low pulsation at flow rate options of 0.001 to 5 mL/min or 0.01 to 10 mL/min. A built-in static mixer ensures reproducible mixing and increased sensitivity. Two head sizes are available for flow rates from 0.001 to10-mL/min to extend separation flexibility.
For more information on the Cole-Parmer LC2 System, go to ColeParmer.com/21019 or call 800-323-4340.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, Digi-Sense™, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at ColeParmer.com.
Download 300 DPI 42662_51.jpg Image