Save time and energy with a low-maintenance alternative to water baths
Vernon Hills, Ill., June 19, 2013—For an eco-friendly selection for labs and research facilities, Cole-Parmer offers Waterless Bead Baths using metallic beads instead of water. The beads are recyclable, use 50% less electricity, and do not require germicides. Yet, the bead bath delivers exceptional temperature uniformity of +/-1 degree and, because the bath is always ready unlike a water bath, no warm-up time is necessary.
The beads create a dry bath, which eliminates the contamination and maintenance issues related to traditional water baths. They also do not require germicides. The beads hold a variety of containers in place without the need for racks or weights. The beads are moisture- and gas-impermeable. Dry, inert, nontoxic aluminum oxide ensures hazard-free operation, including emptying, cleaning, and refilling.
To view the Waterless Bead Baths, go to ColeParmer.com/20139 or call 800-323-4340 for more information.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at ColeParmer.com
Download 300 DPI 10122_00.jpg Image