Cole-Parmer Announces Exclusive Partnership with picoSpin™, LLC
3/2/2011
With the launch of the world’s first miniature nuclear magnetic resonance (NMR) spectrometer, Cole-Parmer is chosen as the exclusive international distribution partner
Vernon Hills, Ill., March 1, 2011—As a revolutionary new breakthrough in spectrometry enters the market, its creators at picoSpin, LLC have engaged Cole-Parmer to sell their product internationally. These exclusive rights apply to the picoSpin™ Benchtop NMR Spectrometer, an innovative and disruptive technology that combines low price, compact footprint, with true spectroscopic capability. With this combination of sought-after features, research labs and educators now have affordable access to a dedicated NMR spectrometer.
“NMR is the most powerful analytical tool in chemistry,” said John Price, Ph.D., president and chief scientist of picoSpin, LLC, “You can now have an NMR system right at your lab bench. You can deploy multiple units within a factory to continuously monitor process fluids and control them all from a web browser anywhere in the world. Your students can experience NMR hands-on in chemistry lab courses and in their research projects.”
The new device is a complete liquid-phase proton NMR system, including permanent magnet, transmitter, receiver, data acquisition, programmable pulse sequencer, Ethernet interface and intuitive web-based control software. As a breakthrough technology, it has already garnered the attention of the scientific community. This month, the picoSpin-45 NMR Spectrometer was named a finalist in the Science & Medical – Diagnostic Aids category, one of twelve categories honored by the Edison Awards.
To view this technology, available exclusively from Cole-Parmer, go to ColeParmer.com/8333 or, for more information, call 847-549-7600.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the U.S., call 800-323-4340. International customers, call 847-549-7600. Visit us at www.coleparmer.com.
Download 300 DPI 83090_00.jpg Image