Lighter, more rugged, and easier to use
Vernon Hills, Ill., March 5, 2012—Spend less time finding problems and more time solving them with the innovative, rugged, and easy-to-use Fluke Ti110/TiR110 and Ti125/TiR125 Thermal Imagers. Their exclusive IR-OptiFlex™ focus-free system ensures that images are well-focused from four feet and beyond for optimal clarity and scanning. The IR-PhotoNotes™ annotation system helps users quickly identify and track inspection locations by adding digital images of critical information. Fluke''s exclusive IR-Fusion® with AutoBlend™ mode pinpoints potential issues by fusing digital and IR images into a single information-filled image.
Choose from two types of advanced thermal imagers. The Industrial-Commercial Thermal Imagers (Ti110 and Ti125) are the best choice for industrial, mechanical, electromechanical, and general maintenance. They also work well for process, refractory insulation, tank and vessel levels, steam systems and traps, pipes and valves, along with electrical, unbalanced loads, overloaded systems, wiring problems or component failure. The Building Diagnostic Thermal Imagers (TiR110 and TiR125) are excellent for building problems, defects, energy audits, building inspection, and weatherization plus restoration, water damage and roofing.
Work faster and more efficiently to document findings with the all-new advanced Fluke Thermal Imagers. For more information, go to www.davis.com/19084 or call 800-358-5525.
Davis Instruments has been one of the world''s leading sources of test, measurement, control, and calibration instruments since 1912. With expanding capabilities in industrial spheres such as power and utilities, petrochemicals, aerospace and electronics production, field services, and construction, the organization continues to respond to technological demands. Based in Vernon Hills, Illinois, Davis Instruments is dedicated to providing customers with excellent service and product applications expertise. For more information, contact Davis Instruments, 625 East Bunker Court, Vernon Hills, IL 60061. In the USA or Canada, call 800-358-5525. International customers, call 847-327-2000. Visit us at www.davis.com.
Download 300 DPI DI_Fluke_Ti110&125Thermal-Imagers.jpg Image