All New! Non-DEHP Lab Tubing from Cole-Parmer
9/5/2012
Nontoxic formulation is safe for contamination-free applications
Vernon Hills, Ill., September 4, 2012—The new Cole-Parmer® Non-DEHP Lab Tubing offers a versatile option for analytical instruments, food processing, and general laboratory uses. The tubing transfers liquids, gases, and inorganic chemicals found in the lab, paint, pigment, and soap dispensing applications. Because it contains no DEHP (di(2-ethylhexyl)phthalate), it is free from leachates that may compromise the transferred material.
Made of clear material, this tubing can be visibly inspected for flow monitoring. It is flexible, with a small bend radius, and installs easily on barbed fittings. This autoclavable tubing meets FDA CFR 21, Part 175.300 certification and is Proposition 65- and REACH-compliant. Use the Cole-Parmer Non-DEHP Lab Tubing for temperatures ranging from -58 to 165°F (-50 to 73°C).
For more information, call 800-323-4340 or visit ColeParmer.com/19444.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at www.coleparmer.com.