New Economical StableTemp Utility Baths Offering Digital Technology Introduced by Cole-Parmer
The new Cole-Parmer StableTemp Economical Digital Utility Water Baths, available in five reservoir sizes, combine precise temperature control and excellent temperature stability to provide reliable heating.
Despite the economical price point, the digital water baths with lid include features such as LCD readings in Fahrenheit or Celsius, easy touch-button recall, built-in timer, and high- temperature alarm and safety shutdown.
These laboratory water baths contain dual thermostats: a primary microprocessor-based temperature controller that delivers +/-0.1°C precision and a secondary overtemperature safety to prevent samples from overheating. For optimal temperature accuracy, the primary controller can be calibrated to a reference thermometer.
The seamless stainless steel tank contains no welds to leak or rust—and the rounded corners provide an easy-to- clean interior. A hinged polycarbonate lid allows condensation to flow back into bath.
To view the Cole-Parmer StableTemp Economy Utility Baths, available in 2-L, 5-L, 10- L, 20-L and 28-L reservoir sizes, go to ColeParmer.co.uk/21346 or, for more information, email [email protected] or call 020-8574-7556.
Download 300 DPI 14576_00_02.jpg Image