TraceableOne™ Single-Use USB Temperature Data Loggers
Automatically generates PDF data report via USB port
  • LED alarm indicator for visual status indication
  • Time delay of 30 minutes before logging begins for enhanced logging accuracy
  • Protective plastic pouch maintains waterproof IP67 rating
  • Bar code for in-house scanning
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
These data loggers can be used for monitoring and recording temperatures during shipments, storage, or experiments. They are ideal for food, pharmaceutical, or laboratory use—any application where a small accurate datalogger is required.
The data logger features a time delay of 30 minutes—so the logger will not alert when the temperature is within the alarm zone, until the delay time elapses. Protective plastic pouch keeps logger dry, eliminating concerns when shipping materials with cold packs, ice, or in damp environments. To maintain IP67 waterproof rating, do not tear open pouch before data download.
Products have an individually-numbered Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Related Article: How to Safely Transport Vaccines, how to meet the critical challenge of ensuring a temperature-controlled supply chain from manufacturer to end-user.
41,200.00 - ₹146,000.00INR / Pkg of 40
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
COMPARE
8 2 Single-Use USB Temperature Data Logger, 10 Day, 1 Minute Interval, 2 to 8˚C; 40/pk
₹133,000.00INR / Pkg of 40
COMPARE
-15 -20 Single-Use USB Temperature Data Logger, 10 Day, 1 Minute Interval, -20 to -15˚C; 40/pk
₹146,000.00INR / Pkg of 40
COMPARE
8 2 Single-Use USB Temperature Data Logger, 30 Day, 3 Minute Interval, 2 to 8˚C; 40/pk
₹133,000.00INR / Pkg of 40
COMPARE
8 2 Single-Use USB Temperature Data Logger, 60 Day, 6 Minute Interval, 2 to 8˚C; 40/pk
₹133,000.00INR / Pkg of 40
COMPARE
-15 -20 Single-Use USB Temperature Data Logger, 60 Day, 6 Minute Interval, -20 to -15˚C; 40/pk
₹133,000.00INR / Pkg of 40
COMPARE
8 2 Single-Use USB Temperature Data Logger, 90 Day, 10 Minute Interval, 2 to 8˚C; 40/pk
₹112,000.00INR / Pkg of 40
COMPARE
-15 -20 Single-Use USB Temperature Data Logger, 90 Day, 10 Minute Interval, -20 to -15˚C; 40/pk
₹133,000.00INR / Pkg of 40
COMPARE
8 2 Single-Use USB Temperature Data Logger, 10 Day, 1 Minute Interval, 2 to 8˚C; 10/pk
₹41,200.00INR / Pkg of 10
Showing 1- 8 of 8