Cole-Parmer® INC-200 Series Stuart Hybridization Incubator/Shakers
Versatile—features both rotisserie and rocking motions
  • Forced air circulation combined with microprocessor control result in excellent temperature uniformity
  • Up-and-over door for easy access to the chamber
Use a hybridization incubator when it is vital to maintain the integrity of the sample while heating it to the exact temperature. The incubator is ideal for nucleic acid hybridization applications such as Northern (RNA) blots, Southern (DNA) blots, and Western (protein) blots. This compact incubator comes with two built-in shakers. Rotisserie shaker has a rotational speed from 2 to 10 rpm while the platform shaker has a rocking speed from 5 to 70 oscillations/minute. The gentle rocking action is ideal for the washing stage of most hybridization procedures.
Safety features include an overtemperature protection system, plus the power is cut to the motor if the motor overheats or stalls. Stainless steel interior is easy to clean. Double-glazed smoked acrylic and polycarbonate door provides radiation protection.
Order a rotisserie that accepts 75 mm OD bottles and hybridization bottles separately. Bottles are made from borosilicate glass and are autoclavable.
631,000.00 - ₹642,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Max Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
0.7 80 115
₹642,000.00INR / Each
COMPARE
0.7 80 230
₹631,000.00INR / Each