Cole-Parmer® SamplePrep Ceramic Grinding Cylinders
Angle-cut ends help shear sample matrix for thorough extraction of analyte into the solvent
  • Constructed of durable ceramic
Ceramic grinding media are excellent for mixing samples for extraction methods such as pesticide residue analysis (QuEChERS) or extracting oil from fish and seafood. Typically two cylinders are used per sample vial.
Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
8,500.00 - ₹8,600.00INR / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Cole-Parmer SamplePrep 2183 Ceramic Grinding Cylinders, Large, 3/8"W x 7/8"L; 100/PK 0457557 Mfr # 2183
COMPARE
Ceramic Grinding Cylinders, Large, 3/8"W x 7/8"L; 100/PK
₹8,600.00INR / Pkg of 100
Cole-Parmer SamplePrep 2184 Ceramic Grinding Cylinders, Medium, 5/16"W x 5/8"L; 100/PK 0457558 Mfr # 2184
COMPARE
Ceramic Grinding Cylinders, Medium, 5/16"W x 5/8"L; 100/PK
₹8,600.00INR / Pkg of 100
Cole-Parmer SamplePrep 2185 Ceramic Grinding Cylinders, Small, 5/32"W x 5/16"L; 100/PK 0457559 Mfr # 2185
COMPARE
Ceramic Grinding Cylinders, Small, 5/32"W x 5/16"L; 100/PK
₹8,500.00INR / Pkg of 100