Cole-Parmer® SamplePrep SM-300 ShatterBox® Versatile Ring and Puck Mill
Grind your samples with minimal loss in a soundproof enclosure
  • Stores up to 10 customizable grinding methods.
  • Accommodates various grinding dishes.
  • Industrial-grade modular electronics.
  • Robust IP65 rated industrial interface.
  • Dedicated PLC-based programming.
  • Single-phase power, no separate power supply.
  • Multilingual interface.
  • USB-upgradeable firmware.
  • Automated door opening.
  • Electronically controlled, fully reproducible grinding methods with networking (ethernet) capabilities.
  • Dual-locking safety door.
  • Real-time monitoring of clamping force.
  • Sound-insulated steel cabinet.
  • Lockable caster wheels.
The SM-300 ShatterBox® is a versatile ring and puck mill that can crush and grind many types and sizes of samples with minimal sample loss and increased safety. The versatility of sample containers allows for grinding a wide range of sample amounts (5 g up to 150 g).
Typical applications include: XRF, XRD, ICP, AAS, and IR analyses for rocks, minerals, cement, ores, slag, soil, and ceramics.
Typical samples include: cement, superalloy, granite, glass, sand, slag, ceramics, seeds, grains, pigments, and catalyst supports.
The Cole-Parmer® SM-300 ShatterBox is a versatile ring and puck mill that can crush and grind many types and sizes of samples with minimal sample loss and increased safety. The ShatterBox features an automated dish positioning system and locking system for ease of use and greater flexibility. The ShatterBox pulverizes up to 100 mL in volume (or 150g in weight) of brittle material to analytical fineness. The unit has many advanced features, including a touchscreen with programmable timer, an automated clamp, a safety-interlocking automated door, and horizontal grinding container insertion/removal.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Speed (rpm) Description
Availability Pricing
Cole-Parmer SamplePrep SM-300 ShatterVersatile Box Ring and Puck Mill; 115 VAC, 60 Hz 4101083 Mfr # 8550-115
COMPARE
115 800 to 1200 (1050 for tungsten carbide dish) ShatterBox® Versatile Ring and Puck Mill; 115 VAC, 60 Hz