Cole-Parmer® SamplePrep HG-600 Geno/Grinder® 2010 Tissue Homogenizer and Cell Lyser
Designed for rapid cell disruption, cell lysis and homogenization of plant and animal tissue through bead beating
  • Ideal for high-throughput cell lysis and applications including DNA/RNA extraction, protein extraction, and pesticide residue analysis
  • Programmable touch screen control panel for run time, rate, cycles, and pause time
  • Up to 500 protocols can be saved for recall
  • Designed for vigorous up-and-down shaking of deep-well titer plates, vial sets, and centrifuge tubes
  • Optional Kryo-Tech® Accessories enable cryogenic grinding and preserve temperature sensitive samples for RNA and protein extractions
  • Comes with an adjustable clamp with a quick release button that allows users to secure vials or titer plates of various sizes
  • Typical sample processing time of 1-2 minutes.
The Cole-Parmer SamplePrep HG-600 Geno/Grinder is a high-throughput plant and animal tissue homogenizer and cell lyser designed to simplify and speed up sample processing, delivering faster, more efficient extractions with consistent results. It is specifically designed for rapid cell disruption, cell lysis and tissue homogenization through bead beating.
Typical applications include: tissue homogenization, DNA/RNA research and extraction, cell lysis, pesticide residue extraction, protein and metabolite extraction, biofuel research, and QuEChERS.
Typical samples include: animal tissue, plant tissue, cell cultures, fruit, cannabis, seeds, yeast, and bacteria.
Save time and increase efficiency by processing multiple samples in minutes, eliminating the need for slow, manual grinding methods. The HG-600 Geno/Grinder 2010 is equipped with an adjustable clamp that accommodates a full range of sample vials from 2 mL to 50 mL centrifuge tubes or deep-well titer plates. A full range of pre-loaded sample vials makes setup quick and easy.
User-friendly and easy to operate touchscreen can be tilted for better viewing. Controls allow you to program and save up to 500 protocols, ensuring repeatability. Protocols can be easily labeled, saved, recalled, and exported via USB. The unit is equipped with an administrative lockout (passcode) which allows you to restrict user access to specific protocols. The double-locking lid provides safe operation, the sound insulation the suspension system ensures quiet, vibration-free operation.
A large observation window and interior LED lighting improves visibility and simplifies monitoring. The quick-release adjustable clamp makes loading and unloading vials or titer plates hassle-free.
The vertically oscillating motion of the unit is designed to effectively disrupt cellular materials enabling the unit to be used to prepare sample tissue for extractions of nucleic acid, proteins, and other constituents by agitating the tissue, steel balls or beads and a lysis agent together in each well of the titer plate, tube or vial.
The Cole-Parmer® SamplePrep Geno/Grinder handles a variety of sample types including seeds, stems, roots, leaves, and animal tissue. The samples can be processed with or without reagents in approximately 60 seconds or less. Optional Kryo-Tech® accessories keep temperature-sensitive samples stable.
With its new design and upgraded features, the HG-600 Geno/Grinder® 2010 is built to make sample preparation easier, faster, and more reliable.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Sample Size (mL) Min Sample Size (mL) Power (VAC)
Availability Pricing
Cole-Parmer SamplePrep HG-600-115 Geno/Grinder® 2010 Tissue Homogenizer and Cell Lyser; 115 VAC, 60 Hz 0450017 Mfr # 2010-115
COMPARE
50 2 115
Call For Price
Cole-Parmer SamplePrep HG-600-230 Geno/Grinder® 2010 Tissue Homogenizer and Cell Lyser; 230 VAC, 50 Hz 0457684 Mfr # 2010-230
COMPARE
50 2 230