Raven Environmental F-10300 Benchtop Centrifuges
Get true, accurate measurements of solids concentration
  • Horizontal 6-place rotor
  • Clarify liquid samples prior to lab testing
Heavy-duty stainless steel and aluminum construction makes this benchtop centrifuge ideal for industrial environments such as food and beverage, wastewater plants, meat packing, and paper mills. Includes six polycarbonate reusable tubes clearly marked to indicate percent volume. The conical shape of the tubes accentuate small amounts of solids concentration to as low as 0.2%.
Separate solids from liquid for improved laboratory tests.
565,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RCF x g Max Speed (rpm) Power (VAC)
Availability Pricing
Raven Environmental F-10300 Benchtop Centrifuge; 230 VAC, 50 Hz 0547715 Mfr # F-10300-230
COMPARE
1792 3000 230
₹565,000.00INR / Each