PolarSafe® EVA Ice Buckets
Mess-free superior insulation for any cryogenic medium
  • Ethylene vinyl acetate is leak proof and will not "sweat"
  • Thick, nontoxic foam is compatible with a variety of cryogenic media
  • Ergonomic handles on bases and lids are glove friendly
  • Pour spouts make cleanup easy
These EVA foam buckets are ideally suited to benchtop chilling applications. They will not create puddles of condensed atmospheric vapor or leak cryogenic media onto your workspace. Ethylene vinyl acetate is compatible with ice, dry ice, liquid nitrogen, alcohol and saline solutions. Buckets feature useful handles and convenient pour spouts for easy use and cleanup, even while wearing cryogenic gloves. Stackable construction ensures compact storage.
9,400.00 - ₹25,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Diameter (OD) (in) Capacity Color
Availability Pricing
COMPARE
7 2.5 L Blue
₹9,400.00INR / Each
COMPARE
7 2.5 L Green
₹9,400.00INR / Each
COMPARE
7 2.5 L Purple
₹9,400.00INR / Each
COMPARE
7 4 L Green
₹25,000.00INR / Each
COMPARE
7 4 L Purple
₹10,500.00INR / Each
COMPARE
7 4 L Black
₹20,000.00INR / Each