Diba Omnifit® EZ Chromatography Columns with Two Adjustable Endpieces
Ergonomically designed endpieces provide simple and quick height adjustment
  • PEEK™ column threads offer outstanding durability
  • Precision borosilicate glass columns provide inertness, visibility, and performance
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Ergonomically designed EZ columns are easy to use yet provide excellent results. Columns are ideal for low- to medium-pressure applications and are resistant to many solvents used in liquid chromatography. Modular design gives you the flexibility to mix and match column sizes with different endpiece types to meet your specific requirements.
Two adjustable endpieces (plungers) provide maximum flexibility in bed height. With an adjustable endpiece at both ends of the column, you gain 160 mm of adjustment plus the ability to make fine adjustments with minimal disturbance to the packed bed. Each glass column assembly is supplied preassembled with 25 μm PE frits and comes with all the tubing, fittings, and connections you need to get started.
64,500.00 - ₹236,000.00INR / Each
44 variations of this product are available.
See All Product Options Below
44 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Max Pressure (PSI)
Availability Pricing
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 5 mm ID x 50 mm L WW-11942-72 Mfr # 006EZ-05-05-AA
COMPARE
5 50 1200
₹123,000.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 5 mm ID x 100 mm L WW-11942-75 Mfr # 006EZ-05-10-AA
COMPARE
5 100 1200
₹123,000.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 5 mm ID x 150 mm L WW-11942-78 Mfr # 006EZ-05-15-AA
COMPARE
5 150 1200
₹123,000.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 5 mm ID x 250 mm L WW-11942-81 Mfr # 006EZ-05-25-AA
COMPARE
5 250 1200
₹125,000.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 5 mm ID x 330 mm L WW-11942-84 Mfr # 006EZ-05-33-AA
COMPARE
5 330 1200
₹132,000.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 5 mm ID x 400 mm L WW-11942-87 Mfr # 006EZ-05-40-AA
COMPARE
5 400 1200
₹146,000.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 6.6 mm ID x 50 mm L WW-11942-90 Mfr # 006EZ-06-05-AA
COMPARE
6.6 50 900
₹64,500.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 6.6 mm ID x 100 mm L WW-11942-93 Mfr # 006EZ-06-10-AA
COMPARE
6.6 100 900
₹72,800.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 6.6 mm ID x 150 mm L WW-11942-96 Mfr # 006EZ-06-15-AA
COMPARE
6.6 150 900
₹67,400.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 6.6 mm ID x 250 mm L WW-11942-99 Mfr # 006EZ-06-25-AA
COMPARE
6.6 250 900
₹67,900.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 6.6 mm ID x 330 mm L WW-11943-02 Mfr # 006EZ-06-33-AA
COMPARE
6.6 330 900
₹70,800.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 6.6 mm ID x 400 mm L WW-11943-05 Mfr # 006EZ-06-40-AA
COMPARE
6.6 400 900
₹70,300.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 10 mm ID x 100 mm L WW-11943-08 Mfr # 006EZ-10-10-AA
COMPARE
10 100 600
₹81,500.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 10 mm ID x 150 mm L WW-11943-11 Mfr # 006EZ-10-15-AA
COMPARE
10 150 600
₹81,900.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 10 mm ID x 250 mm L WW-11943-14 Mfr # 006EZ-10-25-AA
COMPARE
10 250 600
₹82,400.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 10 mm ID x 330 mm L WW-11943-17 Mfr # 006EZ-10-33-AA
COMPARE
10 330 600
₹84,000.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 10 mm ID x 400 mm L WW-11943-20 Mfr # 006EZ-10-40-AA
COMPARE
10 400 600
₹85,200.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 10 mm ID x 500 mm L WW-11943-23 Mfr # 006EZ-10-50-AA
COMPARE
10 500 600
₹88,600.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 15 mm ID x 100 mm L WW-11943-26 Mfr # 006EZ-15-10-AA
COMPARE
15 100 300
₹103,000.00INR / Each
Diba Omnifit® EZ Chromatography Column with 2 Adjustable Endpieces, 50 mm ID x 250 mm L WW-11943-74 Mfr # 006EZ-50-25-AA
COMPARE
50 250 75
₹218,000.00INR / Each
Showing 1 - 20 of 44