Environmental Express Oakton® T-100 Handheld Turbidity Meter
Waterproof and dustproof with IP67-rated housing that floats!
  • Meets performance criteria as specified by ISO 7027 (DIN EN 27027) method
  • Compliant with ASTM D6855-03 (EPA recognized)
  • Four point push-button calibration for full-range accuracy
  • Auto-ranging from 0 to 1000 NTU
The Oakton T-100 turbidity meter is easy to calibrate — meter automatically prompts you to the next calibration standard. The large custom LCD displays reading and units of measure. Other features include auto-off, diagnostic error messages, and auto-ranging function. Power saving function lets you take over 1000 readings on a single set of AAA batteries. Sealed optical system with infrared light source delivers high accuracy across entire range.
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
T-100 Turbidity Meter, handheld, waterproof, no standards
₹326,000.00INR / Each