Oakton® pH Buffer Solutions

Ideal for laboratory and field analysis
  • All Oakton pH buffers are traceable to NIST standard reference materials
  • Buffer solution lets you calibrate pH instruments to ensure accuracy
  • If unopened, shelf life is 2 years
• Oakton pH Buffer Solutions have pH/temperature tables printed on the labels for accurate calibrations (not on 60-mL, but included as insert)
• Manufactured under ISO 9000 quality guidelines and traceable to NIST standards
• If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days
• Buffer Options:
- pH 1.68 buffer available in clear
- pH 4.01 buffer available in red
- pH 6.86 buffer available in clear
- pH 7.00 buffer available in yellow
- pH 9.18 buffer available in clear
- pH 10.01 buffer available in blue
- pH 12.46 buffer available in clear
1,300.00 - ₹21,000.00INR / Pint of 1

26 variations of this product are available.

See All Product Options Below
26 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Buffer Value Volume (mL)
Availability Pricing
COMPARE
1.68 60
₹8,100.00INR / Pkg of 5
COMPARE
4.01 60
₹8,100.00INR / Pkg of 5
COMPARE
7 60
₹8,100.00INR / Pkg of 5
COMPARE
10.01 60
₹8,100.00INR / Pkg of 5
COMPARE
6.86 60
₹8,100.00INR / Pkg of 5
COMPARE
9.18 60
₹12,000.00INR / Pkg of 5
COMPARE
12.46 60
₹11,900.00INR / Pkg of 5
COMPARE
4.01 500
₹1,400.00INR / Pint of 1
COMPARE
1.68 500
₹2,800.00INR / Each
COMPARE
6.86 500
₹1,400.00INR / Each
COMPARE
7 500
₹1,500.00INR / Pint of 1
COMPARE
9.18 500
₹1,400.00INR / Each
COMPARE
10.01 500
₹1,400.00INR / Pint of 1
COMPARE
12.46 500
₹1,300.00INR / Each
COMPARE
4.01 500
₹17,100.00INR / Case of 12
COMPARE
4.01 1000
₹2,800.00INR / Each
COMPARE
4.01 4000
₹4,200.00INR / Each
COMPARE
4.01 4000
₹21,000.00INR / Case of 4
COMPARE
7 500
₹17,100.00INR / Case of 12
COMPARE
7 1000
₹2,700.00INR / Each
Showing 1 - 20 of 26

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat