Environmental Express Oakton® PC 700 Benchtop pH/Conductivity Meters
Easily toggle between pH, mV, conductivity, TDS, and temperature measurements at the touch of a button
  • Space-saving design takes up less room on the bench than other meters
  • Large display lets you see your measurements from across the lab
  • Conductivity and TDS autoranging chooses the best resolution for each measurement
The PC700 meter offers a new compact design that is over 40% smaller than other benchtop meters. The large display is easy to read even at a distance. Dual-display allows you to read both a measured parameter and temperature at a glance. The meter features pH auto-buffer recognition for both USA and NIST buffer sets (1.68, 4.01, 7.00, 10.01 and 12.45 or 1.68, 4.01. 6.18. 9.86, and 12.45), automatic or manual temperature compensation, temperature in °C, display Hold, and water-resistant membrane keypad.
Previous calibration points, electrode slope and offset, and conductivity/TDS cell constants (k=0.1, k=0.475 ) are stored in memory for quick and easy retrieval. The convenient pull-out quick reference card keeps keypad functions at your fingertips.
141,000.00 - ₹278,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
NIST Calibration Included Probe Included Description
Availability Pricing
COMPARE
No Yes Meter w/pH electrode, conductivity/temp probe, and electrode stand
₹190,000.00INR / Each
COMPARE
Yes Yes PC 700 Benchtop Meter with Probes and NIST Calibration
₹278,000.00INR / Each
COMPARE
No No PC 700 Benchtop Meter with Electrode Stand
₹141,000.00INR / Each