Oakton® CON 6+ Handheld Conductivity Meters
Autoranging conductivity with temperature measurement
  • CON 6+ meters have five auto-ranging measurement modes — up to 200.0 mS (200.0 ppt)
  • Temperature readout in °C with the push of a button
  • Rubber boot with built-in stand protects meter and adds functionality
These microprocessor-based meters feature easy, push-button operation that offers high accuracy at an extremely affordable price. A quick and easy single-point calibration covers all five ranges with the CON 6+ meters; for greater precision, single points can be calibrated within each of the five measurement ranges.
Additional features include a convenient Hold function and auto-off function that shuts meter off after 20 minutes of nonuse. Programming functions allow customization of meter for temperature compensation, temperature coefficient, normalization, and cell constant for better accuracy in varying applications.
75,600.00 - ₹97,200.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Conductivity Resolution Max Conductivity Min Conductivity
Availability Pricing
COMPARE
0.01, 0.1, or 1 μS; 0.01, or 0.1 mS 0 - 200 mS 0 -2000 uS
₹75,600.00INR / Each
COMPARE
0.01, 0.1 or 1 μS; 0.01, or 0.1 mS 0 - 200 mS 0 - 2000 uS
₹87,100.00INR / Each
COMPARE
0.01 , 0.1, or 1 μS; 0.01, or 0.1 mS 0 - 200 mS 0 - 2000 uS
₹86,200.00INR / Each
COMPARE
0.01, 0.1, or 1 μS; 0.01, or 0.1 mS 0 - 200mS 0 - 2000 uS
₹97,200.00INR / Each