Argos Technologies® Magne-Box Magnetic Polycarbonate Cryo-Boxes
Added security for your cryo samples
  • Boxes fit in standard size freezer racks
  • Polycarbonate design withstands temperatures from -196 to 121°C
  • Slot openings on the bottom of boxes allow for drainage
  • 25-, 81-, and 100-place boxes accommodate 2 mL cryovials
  • 81-Tall boxes accommodate 4-5 mL cryovials
Argos Technologies® Magne-Box™ freezer boxes are the first of its kind to offer a magnetic lid closure! Each box features magnets that securely connect the base to the hinged lid. This allows for one handed retrieval from freezer racks and ensures that samples stay safe. Polycarbonate boxes have clear lids with colored inserts and numbered grids to facilitate easy sample identification. Available with 25-, 81-, and 100-place divisions.
8,100.00 - ₹11,000.00INR / Pkg of 8
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Height (in) Tubes Held
Availability Pricing
COMPARE
Orange 2 1/16 25
₹11,000.00INR / Pkg of 8
COMPARE
Assortment 2 1/16 81
₹8,100.00INR / Pkg of 4
COMPARE
Blue 3 3/4 81
₹9,900.00INR / Pkg of 4
COMPARE
Green 5 1/5 100
₹8,500.00INR / Pkg of 4