Cole-Parmer® TELOS® QuEChERS Cleanup Kits
Optimized to perform more advanced clean up of the sample
  • Remove pigments, acids, and fats using three different combinations of SPE sorbents
  • Anhydrous magnesium sulfate removes any excess water
Further clean up of samples is easy with TELOS QuEChERS clean up kits. After initial sample prep with the extraction kit, a more advanced clean up is often required for the removal of interferences such as pigments, organic acids and fats. The removal of these interferences is carried out using a combination of three different SPE sorbents (see below).
TELOS PestiCarb: removes pigments, including chlorophyll and carotenoids
TELOS PSA: removes organic acids
TELOS C18: removes nonpolar interferences, including fats
18,100.00 - ₹39,100.00INR / Pkg of 100
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (mL) Applications Methodology
Availability Pricing
COMPARE
2 Removal of fats and proteins: 50 mg PSA, & 50 mg C18 AOAC 2007.01
₹22,700.00INR / Pkg of 100
COMPARE
2 Removal of fats and proteins: 25 mg PSA & 25 mg C18 EN 15662
₹27,800.00INR / Pkg of 100
COMPARE
15 Removal of fats and proteins: 400 mg PSA & 400 mg C18 AOAC 2007.01
₹19,700.00INR / Pkg of 50
COMPARE
15 Removal of fats and proteins: 150 mg PSA & 150 mg C18 EN 15662
₹27,800.00INR / Pkg of 50
COMPARE
2 Romoval of fats, proteins, and pigments: 50 mg PSA, 50 mg C18, & 50 mg PestiCarb AOAC 2007.01
₹39,100.00INR / Pkg of 100
COMPARE
2 General cleanup: 50 mg PSA AOAC 2007.01
₹21,300.00INR / Pkg of 100
COMPARE
2 General cleanup: 25 mg PSA EN 15662
₹22,100.00INR / Pkg of 100
COMPARE
15 General cleanup: 150 mg PSA EN 15662
₹27,800.00INR / Pkg of 50
COMPARE
15 General cleanup: 400 mg PSA AOAC 2007.01
₹18,100.00INR / Pkg of 50
COMPARE
2 Removal of pigments: 50 mg PSA & 50 mg PestiCarb AOAC 2007.01
₹35,500.00INR / Pkg of 100
COMPARE
2 Removal of pigments: 25 mg PSA & 2.5 mg PestiCarb EN 15662
₹27,300.00INR / Pkg of 100
COMPARE
2 Removal of pigments: 25 mg PSA & 7.5 mg PestiCarb EN 15662
₹34,300.00INR / Pkg of 100
Showing 1- 12 of 12