International Products Corp LF2100® Low-Foam Cleaner, Liquid Detergent

LF2100® liquid low-foam cleaner
  • Ideal for automatic washers Available in 1-L bottles to 235 kg plastic drums!
A mild yet powerful cleaner for automatic washing machines and other high-pressure/high-agitation applications. Removes contaminants such as grease, oil, wax, particulates, tar, ink and most other soils from glass, stainless steel, ceramics, plastics, and many other surfaces. It is effective at 1 – 2% in water and rinses clean, leaving no residue. Meets USDA guidelines for incidental contact with food processing systems. Solution is safe for workers and the environment – contains no phosphates, CFCs, ODCs, phenols, or borates.

Note: May etch zinc, magnesium, and aluminum.
8,700.00 - ₹123,000.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Format Description
Availability Pricing
International Products Corp LF2100® Low-Foam Cleaner, Liquid Detergent; 235 kg 1810006 Mfr # L-2190-055
COMPARE
Liquid LF2100® Low-Foam Cleaner, Liquid Detergent; 235 kg
Call For Price
International Products Corp LF2100® Low-Foam Cleaner, Liquid Detergent; 1 Liter 1810200 Mfr # L-2101-12
COMPARE
Liquid LF2100® Low-Foam Cleaner, Liquid Detergent; 1 Liter
₹8,700.00INR / Each
International Products Corp LF2100® Low-Foam Cleaner, Liquid Detergent; 12 x 1 L 1810210 Mfr # L-2101-12
COMPARE
Liquid LF2100® Low-Foam Cleaner, Liquid Detergent; 12 x 1 L
₹94,100.00INR / Case of 12
International Products Corp LF2100® Low-Foam Cleaner, Liquid Detergent; 20 L 1810230 Mfr # L-2120
COMPARE
Liquid LF2100® Low-Foam Cleaner, Liquid Detergent; 20 L
₹123,000.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat