Cole-Parmer® HP-200 Series Undergrad Analog Hot Plates
Get accurate surface temperature in real time
  • Compact, stackable design takes up less benchtop and storage space
  • Hot warning light remains on even when unplugged
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
  • Built with tough aluminum casing for longevity
  • Designed to prevent accidental spillage on control panel
Designed with microprocessor-controlled dual thermocouples, these hot plates offer you accurate temperature control and protection from overheating. A colored LED indicator surrounds the heat control knob illuminating the actual surface temperature in real time. The square plate design allows you flexibility of vessel sizes and combinations. Up to four-250 mL beakers can easily fit on the surface at the same time.
To keep you safe, a “hot” warning light flashes whenever the plate surface temperature is above 50 °C and continues to flash until the temperature drops below 50 °C, even when unplugged.
The hot plates feature BioCote® antimicrobial protection which reduces the presence of odor and stain causing microbes such as bacteria, mold, and fungi on the product surface by up to 99.99%.
All models include an integral fitting for a retort rod and allow the retort base to slide underneath. For convenience and power saving, the units are built with a standard mains switch. To achieve accurate sample temperature control up to 200 °C (392 °F), a temperature controller (sold separately) can be connected to the unit.
49,400.00 - ₹55,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Top Plate Material
Availability Pricing
COMPARE
450 230 Ceramic
₹50,200.00INR / Each
COMPARE
450 120 Ceramic
₹55,900.00INR / Each
COMPARE
325 230 Coated Aluminum
₹49,400.00INR / Each
COMPARE
325 120 Coated Aluminum
₹50,900.00INR / Each