Breakthrough, automated processing from bulk to test portions in 15 minutes or less
- High-throughput processing of up to 1 kg of sample per run.
- Capability to use liquid nitrogen or dry ice to process your sample
- Unique and patented technology with automated dispensing of liquid nitrogen eliminates the handling and manipulation of liquid nitrogen
- Intuitve LCD screen interface
- Automatic dispensing of liquid nitrogen
- Complete a method in 15 minutes or less
- High-throughput processing up to 1 kg
The CG-900 Cryo-Blade™ revolutionizes bulk sample processing, delivering finely ground, homogeneous test portions in 15 minutes or less. Designed for efficiency, safety, and convenience, this high-capacity cryogenic grinder handles up to 1 kg of sample per run, making it an essential tool for laboratories needing fast, repeatable, and contamination-free sample preparation.
What’s included: Cryo-Blade unit, insulated outer bowl, inner bowl, cryogenic processor lid with blade USB memory stick, and instruction manual.
Typical applications include: analysis of organic pollutants in food products (pesticide residues), QuEChERS. The unit accommodates an extensive range of both dry and wet samples from watermelon to the toughest of samples such as muscle tissue.
Typical sample include: fruits, vegetables, seeds, fungi, and animal tissue.
The Cole-Parmer SamplePrep CG-900 Cryo-Blade allows you to process more samples in less time. The unit handles up to 1 kg per run, significantly reducing prep time. Highly uniform results are delivered, just 1 gram is needed as a representative test sample. The high-throughput capability saves valuable time for your laboratory and meets Food and Drug Administration (FDA) regulations.
The CG-900 includes safe, controlled, automated dispensing of liquid nitrogen directly onto the sample, minimizing manual handling and ensuring consistent cooling. This innovative technology eliminates the manual handling risks associated with liquid nitrogen and provides the user a liquid nitrogen contact-free experience. With automated liquid nitrogen dispensing, the sample reaches cryogenic temperatures and then activates a grinding phase to pulverize samples, resulting in consistent process control and repeatable results, every time. The unit can also operate with dry ice when liquid nitrogen is unavailable or not required.
Automatic and programmable modes let you choose pre-set parameters or customize up to ten saved methods. The intuitive touchscreen interface includes 12 language options for effortless navigation.
Innovative cryogenic grinding technology uses a spinning blade to efficiently grind frozen samples into a fine, consistent powder. The Cole-Parmer SamplePrep Cryo-Blade maintains optimal cryogenic temperatures, ensuring even challenging materials are properly processed.
The Cryo-Blade is designed for safety and convenience. The insulated outer bowl keeps surfaces safe to touch at all times (although it is recommended to use protective gloves). The removable 5 L inner bowl wallows for easy cleaning, sample storage in the freezer, or quick batch switching giving the user the option of using a second bowl to prepare a new batch. The built-in exhaust system efficiently removes excess gas, keeping the workspace safe. The exhaust can be directly connected to the building’s main exhaust system or placed under a vent hood.
The Cole-Parmer SamplePrep CG-900 Cryo-Blade is built for performance and repeatability. The smart liquid nitrogen dispensing adjusts cooling based on sample size, optimizing efficiency. The temperature monitoring system ensures samples stay at the ideal cryogenic conditions for grinding. Load cell technology measures sample weight, ensuring the precise amount of liquid nitrogen is used for each run.