The product you searched is discontinued. Below is the suggested replacement. Compare to discontinued item.
Thermo Scientific 9106BNWP pH Probe, Sealed/SJ/Epoxy; WP BNC
Representative image only.
Thermo Scientific 9106BNWP pH Probe, Sealed/SJ/Epoxy; WP BNC
Thermo Scientific | Mfr: 9106BNWP| Item: 0571241
Value-priced, durable and low maintenance
₹31,930.00INR / Each
₹31,930.00INR / Each
Product Description
Key Features
  • Low cost epoxy body
  • 1 year warranty
More About this Item
Measure the pH of virtually any sample. Sealed, gel-filled design means there is little electrode maintenance to perform. Single-junction reference means electrodes are suited to most general applications.
1 Year Warranty
Specifications
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionWick
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)0
  • Max pH (pH)12
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC (waterproof when used with Thermo Scientific Orion Star Series meters)
  • Cable Length (m)1
  • DescriptionpH Probe, Sealed/SJ/Epoxy; WP BNC