Cole-Parmer Fluidized Sand Bath Compressed Air Filter
Representative image only.
Cole-Parmer Fluidized Sand Bath Compressed Air Filter
Cole-Parmer | Item: 0119101
Call For Price
Call For Price
Buy It Together
Current Item
Cole-Parmer Fluidized Sand Bath Compressed Air Filter
Cole-Parmer Fluidized Sand Bath Compressed Air Filter
0119101
Call For Price
+ + + =
Total Price 679000.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • 6 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Parts and accessories to meet your application needs and keep your sand bath operating dependably.
Air supply to fluidized sand bath must be clean to preserve bath internal components.
Specifications
  • Compatible WithFluidized Sand Baths
  • DescriptionFluidized Sand Bath Compressed Air Filter