Cole-Parmer Fluidized Sand Bath Air Pressure Regulator with Gauge
Representative image only.
Cole-Parmer Fluidized Sand Bath Air Pressure Regulator with Gauge
Cole-Parmer | Item: 0119102
Call For Price
Call For Price
Buy It Together
Current Item
Cole-Parmer Fluidized Sand Bath Air Pressure Regulator with Gauge
Cole-Parmer Fluidized Sand Bath Air Pressure Regulator with Gauge
0119102
Call For Price
+ + + =
Total Price 679000.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • 6 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Parts and accessories to meet your application needs and keep your sand bath operating dependably.
Air supply to fluidized sand baths must be regulated to 3 psi and 3 cu ft per minutes for proper bath function.
Specifications
  • Compatible WithFluidized Sand Baths
  • DescriptionFluidized Sand Bath Air Pressure Regulator with Gauge