The product you searched is discontinued. Below is the suggested replacement. Compare to discontinued item.
SHOWA 874-08 Gloves, smooth, medium
Representative image only.
SHOWA 874-08 Gloves, smooth, medium
SHOWA | Mfr: 874-08| Item: 8635701
Perfect for chemical handling, spill control, and hazmat applications
₹10,900.00INR / Pair of 1
₹10,900.00INR / Pair of 1
Buy It Together
Current Item
SHOWA 874-08 Gloves, smooth, medium
SHOWA 874-08 Gloves, smooth, medium
8635701
₹10,900.00INR / Pair of 1
+
SHOWA 892-11 Viton Gloves, extra-large, 12-mil thickness, 1 pair
8628823
₹34,400.00INR / Pair of 1
=
Total Price 45300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These gloves provide excellent chemical resistance to hazardous materials — gases, aldehydes, amines, and ketones — without being bulky or restrictive. They are unlined to ensure an excellent grip. Gloves are 14" long with rolled cuffs that fit all hazmat suits.
Compliance: ASTM F-739
Specifications
  • Thickness (mil)14
  • MaterialButyl
  • SizeMedium
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • GripSmooth
  • CuffRolled
  • ComplianceASTM F739
  • SterileNo
  • DescriptionGloves, smooth, medium