The product you searched is discontinued. Below is the suggested replacement. Compare to discontinued item.
Scienceware F371350000 Spinvane Stirring Bar, PTFE, Triangular, 3 mL
Representative image only.
Scienceware F371350000 Spinvane Stirring Bar, PTFE, Triangular, 3 mL
Scienceware | Mfr: F371350000| Item: 0477534
₹2,800.00INR / Each
₹2,800.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
This stir bar is designed for test tubes, micro vials and conical bottom centrifuge tubes.
1 Year Warranty
Specifications
  • Max Stirring Volume (Liters)0.003
  • DescriptionSpinvane Stirring Bar, PTFE, Triangular, 3 mL