Raven Environmental SID-20200-30 Environmental Products Sludge Interface Detector with 30 Foot Cable
Representative image only.
Raven Environmental SID-20200-30 Environmental Products Sludge Interface Detector with 30 Foot Cable
Raven Environmental | Mfr: SID-20200-30| Item: 0595620
Quickly locate sludge blanket depth
₹386,000.00INR / Each
₹386,000.00INR / Each
Buy It Together
Current Item
Raven Environmental SID-20200-30 Environmental Products Sludge Interface Detector with 30 Foot Cable
Raven Environmental SID-20200-30 Environmental Products Sludge Interface Detector with 30 Foot Cable
0595620
₹386,000.00INR / Each
+ =
Total Price 540000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Audio and visal LED indicators
  • Sensitivity adjustment
  • Low battery indicator
  • 1 year warranty
More About this Item
The SID-10200 is a portable handheld instrument that accurately determines the sludge blanket solid/liquid interface depths in primary tanks, clarifiers, sedimentation basins and a wide variety of sludge settling tanks.
The solid/liquid interface is detected when the probe is lowered into the tank. As solids density increses the meter's LED lightbars begin to illuminate and the alarm horn frequency increases. The meter has a sensor with a 30-ft cable that has 1-ft incremented depth marks. The unit is powered by six AA batteries (included) and measures 8"L x 4.5"W x 3.5"D
Related Article: What is Wastewater?
1 Year Warranty
Specifications
  • Power6x AA batteries (included)
  • DescriptionEnvironmental Products Sludge Interface Detector with 30 Foot Cable