Environmental Express pH Electrode, Combination, Double-Junction, Gel Filled
Representative image only.
Environmental Express pH Electrode, Combination, Double-Junction, Gel Filled
Environmental Express | Item: 5900170
Combine durability with reliability
₹15,000.00INR / Each
₹15,000.00INR / Each
Buy It Together
Current Item
Environmental Express pH Electrode, Combination, Double-Junction, Gel Filled
Environmental Express pH Electrode, Combination, Double-Junction, Gel Filled
5900170
₹15,000.00INR / Each
+
Environmental Express Electrode Storage Solution; 500 mL
0065304
₹3,700.00INR / Pint of 1
+ + + + + + + + =
Total Price 532100.00 ADD 10 ITEMS TO CART
Product Description
Includes
3.3-ft (1-m) cable with BNC connector.
Key Features
  • Epoxy-body housing resists impact
  • Ideal for general purpose lab and field applications
  • Compatible with most brands of pH meters
  • 6 month warranty
More About this Item
The Ag/AgCl combination reference electrodes have an impact-resistant epoxy body. Electrodes provide a fast, stable response and are ideal for prolonged readings. Gel-filled design requires virtually no maintenance. 127 mm L x 12 mm diameter with maximum 176F (80C) temperature.
6 Month Warranty
Specifications
  • ApplicationspH
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Max Temperature (° C)80
  • ConnectorBNC
  • DescriptionpH electrode, combination, double-junction, gel filled
  • Warranty6 month warranty